2,3,5-三碘苯甲酸 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
TIBA
T0451-25G
T0451-5G
TCIAT0451-25GEA
0
CNY
TCIAT0451-25G
TCIAT0451-5G
2,3,5-三碘苯甲酸 ≥98.0% (by HPLC, titration analysis)
2,3,5-三碘苯甲酸
分子式:
C₇H₃I₃O₂ 分子量: 499.81 g/mol 沸点: 416 溶点: 225.9 存储条件: Freezer |
MDL编号:
MFCD00002420 CAS号: 88-82-4 EINECS: 201-859-6 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 227.0 to 234.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...