5-碘香兰素 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
4-Hydroxy-3-iodo-5-methoxybenzaldehyde
I0427-25G
I0427-5G
TCIAI0427-25GEA
0
CNY
TCIAI0427-25G
TCIAI0427-5G
5-碘香兰素 ≥98.0% (by GC)
5-碘香兰素
分子式:
C₈H₇IO₃ 沸点: 335 °C (1013 hPa) 溶点: 186…187 °C |
MDL编号:
MFCD00006941 CAS号: 5438-36-8 EINECS: 226-617-7 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 181.0 to 184.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...