五氟苯酚 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
2,3,4,5,6-五氟苯酚
P0919-10G
P0919-25G
TCIAP0919-10GEA
0
CNY
TCIAP0919-10G
TCIAP0919-25G
五氟苯酚 ≥98.0% (by GC, titration analysis)
五氟苯酚
分子式:
C₆HF₅O 沸点: 143°C 溶点: 34…36 °C 密度: 1,75 g/cm³ (20 °C) 存储条件: Ambient |
MDL编号:
MFCD00002156 CAS号: 771-61-9 EINECS: 212-235-8 ADR: 8,II |
|
产品技术规格
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...