4-氟苯酚 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

F0160-25G F0160-500G
TCIAF0160-25GEA 0 CNY
TCIAF0160-25G TCIAF0160-500G
4-氟苯酚 ≥99.0% (by GC)
4-氟苯酚
分子式: C₆H₅FO
分子量: 112.1 g/mol
沸点: 185°C
闪点: 68
存储条件: Refrigerator
MDL编号: MFCD00002316
CAS号: 371-41-5
EINECS: 206-736-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1960455824ec284d185642fd05aad078e967b2dc&vid=e2068d4550fbf4f890f2cc6ad8869b6088840f12&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR