Pentafluoroanisole ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,3,4,5,6-Pentafluoroanisole

P0918-25G P0918-5G
TCIAP0918-25GEA 0 CNY
TCIAP0918-25G TCIAP0918-5G
Pentafluoroanisole ≥98.0% (by GC)
Pentafluoroanisole
分子式: C₇H₃F₅O
沸点: 138°C
溶点: –37 °C
密度: 1,496 g/cm³ (20 °C)
闪点: 32°C
MDL编号: MFCD00000289
CAS号: 389-40-2
EINECS: 206-866-8
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=846ce2d91a89412e90cbacb78e1eb14067579c2d&vid=db2bde2da5ea4927e2e82b56c0f500b8156d5422&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR