五氟硝基苯 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1,2,3,4,5-Pentafluoro-6-nitrobenzene
P1228-25G
P1228-5G
TCIAP1228-25GEA
0
CNY
TCIAP1228-25G
TCIAP1228-5G
五氟硝基苯 ≥97.0% (by GC)
五氟硝基苯
分子式:
C₆F₅NO₂ 沸点: 160°C 闪点: 91°C |
MDL编号:
MFCD00007049 CAS号: 880-78-4 EINECS: 212-915-4 |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...