1-氟-2-硝基苯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: o-Fluoronitrobenzene, 2-Fluoronitrobenzene

F0104-25G F0104-500G
TCIAF0104-25GEA 0 CNY
TCIAF0104-25G TCIAF0104-500G
1-氟-2-硝基苯 ≥98.0% (by GC)
1-氟-2-硝基苯
分子式: C₆H₄FNO₂
分子量: 141.1 g/mol
沸点: 222°C
溶点: -8
闪点: 94°C
存储条件: Ambient
MDL编号: MFCD00007048
CAS号: 1493-27-2
EINECS: 216-088-0
UN号: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=695fd6664abbcc2654ea2319e3a60baba07a8d18&vid=e64a1b14ed8954eb815f05c535fc3a40406c2070&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Yellow to Amber to Dark red clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR