2,4-二氟硝基苯 ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: 2,4-二氟-1-硝基苯

D1636-25G D1636-500G
TCIAD1636-25GEA 0 CNY
TCIAD1636-25G TCIAD1636-500G
2,4-二氟硝基苯 ≥99.0% (by GC)
2,4-二氟硝基苯
分子式: C₆H₃F₂NO₂
沸点: 207°C
溶点: 10 °C
密度: 1,4571 g/cm³ (14 °C)
闪点: 90°C
MDL编号: MFCD00007050
CAS号: 446-35-5
EINECS: 207-167-0
UN号: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d44b302a084181b91cbfd64addf24ead58824f2&vid=42b76981eccc32460505687df0a33ba7ec795e13&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR