3,5-二氟苯胺 ≥98.0% (by GC)

供应商: TCI
Danger

D2665-25G D2665-5G
TCIAD2665-25GEA 0 CNY
TCIAD2665-25G TCIAD2665-5G
3,5-二氟苯胺 ≥98.0% (by GC)
3,5-二氟苯胺
分子式: C₆H₅F₂N
分子量: 129.11 g/mol
沸点: 82°C
存储条件: Ambient
MDL编号: MFCD00007763
CAS号: 372-39-4
EINECS: 206-752-8
UN号: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eb16bd465b107166f27a9a34acd01f91641cada&vid=0540b3e5a1e56e494390a1e4ec827bc20f4f982d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Green to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR