4-(三氟甲氧基)苯甲酸 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
T1400-1G
T1400-5G
TCIAT1400-1GEA
0
CNY
TCIAT1400-1G
TCIAT1400-5G
4-(三氟甲氧基)苯甲酸 ≥97.0% (by GC, titration analysis)
4-(三氟甲氧基)苯甲酸
分子式:
C₈H₅F₃O₃ 分子量: 206.12 g/mol 沸点: 272 溶点: 153.4 |
MDL编号:
MFCD00002541 CAS号: 330-12-1 EINECS: 206-352-3 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 152.0 to 155.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...