2-氨基-5-氟苯甲酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-氟邻氨基苯甲酸

F0396-1G F0396-5G
TCIAF0396-1GEA 0 CNY
TCIAF0396-1G TCIAF0396-5G
2-氨基-5-氟苯甲酸 ≥98.0% (by HPLC, titration analysis)
2-氨基-5-氟苯甲酸
分子式: C₇H₆FNO₂
分子量: 155.13 g/mol
沸点: 305
溶点: 186
存储条件: Ambient
MDL编号: MFCD00055566
CAS号: 446-08-2
EINECS: 207-159-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc9a661b20d8d5d414c87bd9c110c49a278359c&vid=3411bf2a66eab313f36f2353bd6c93179a91cc08&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR