2,4,5-三氟苯腈 ≥98.0% (by GC)

供应商: TCI
Danger

T2326-25G T2326-5G
TCIAT2326-25GEA 0 CNY
TCIAT2326-25G TCIAT2326-5G
2,4,5-三氟苯腈 ≥98.0% (by GC)
2,4,5-三氟苯腈
分子式: C₇H₂F₃N
分子量: 157.09 g/mol
闪点: 83°C
MDL编号: MFCD00013289
CAS号: 98349-22-5
UN号: 3276
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5201ba6178268ef6851fe482dfb24b31edc8a668&vid=e40b8b14ff214816436aa9351b61190c7189647c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR