3-氟-4-甲氧基苯甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 3-氟对茴香醛

F0480-1G F0480-5G
TCIAF0480-1GEA 0 CNY
TCIAF0480-1G TCIAF0480-5G
3-氟-4-甲氧基苯甲醛 ≥98.0% (by GC)
3-氟-4-甲氧基苯甲醛
分子式: C₈H₇FO₂
分子量: 154.14 g/mol
沸点: 132°C
MDL编号: MFCD00003349
CAS号: 351-54-2
EINECS: 206-514-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56956807adbd5a53a33a57ae9da3afd039c8b855&vid=ec84ae7d21209f4bb9ab38aeef0e05e84747ec4e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Green powder to lump to clear liquid
Purity (GC) min. 98.0 %
Freezing point 27.0 to 31.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR