2,3,4,5,6-五氟苯甲醛 ≥96.0% (by GC)

供应商: TCI
Danger

Synonyms: 2,3,4,5,6-Pentafluorobenzaldehyde

P0746-25G P0746-5G
TCIAP0746-25GEA 0 CNY
TCIAP0746-25G TCIAP0746-5G
2,3,4,5,6-五氟苯甲醛 ≥96.0% (by GC)
2,3,4,5,6-五氟苯甲醛
分子式: C₇HF₅O
分子量: 196.08 g/mol
沸点: 170°C
溶点: 24
存储条件: Ambient
MDL编号: MFCD00003303
CAS号: 653-37-2
EINECS: 211-502-6
UN号: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d8888ce1989e9adf16fb985fdd013c60f81b212&vid=de239f5bf5f29dfe60bbe254ecd449e75382e810&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR