2,4-二氟苯甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

D2839-25G D2839-5G
TCIAD2839-25GEA 0 CNY
TCIAD2839-25G TCIAD2839-5G
2,4-二氟苯甲醛 ≥98.0% (by GC)
2,4-二氟苯甲醛
分子式: C₇H₄F₂O
分子量: 142.1 g/mol
沸点: 82
密度: 1.299
闪点: 55°C
存储条件: Ambient
MDL编号: MFCD00010326
CAS号: 1550-35-2
EINECS: 216-287-2
UN号: 1989
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69cd863bdfbe5aaedf2120aa8e664fd46f53576f&vid=49885215fb985566ed2ab3c3e6ba38ba26894014&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR