1,2,3,4-四氟苯 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
T2410-25G
T2410-5G
TCIAT2410-25GEA
0
CNY
TCIAT2410-25G
TCIAT2410-5G
1,2,3,4-四氟苯 ≥98.0% (by GC)
1,2,3,4-四氟苯
分子式:
C₆H₂F₄ 分子量: 150.08 g/mol 沸点: 94°C 溶点: –42°C 密度: 1.811 闪点: 20°C |
MDL编号:
MFCD00000285 CAS号: 551-62-2 EINECS: 208-997-6 UN号: 1993 ADR: 3 II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |