2,4,5-Trichlorothiophenol ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 1-Mercapto-2,4,5-trichlorobenzene, 2,4,5-Trichlorothiophenol, 2,4,5-Trichlorobenzenethiol, 2,4,5-Trichlorophenyl Mercaptan

T0728-10G T0728-25G
TCIAT0728-10GEA 0 CNY
TCIAT0728-10G TCIAT0728-25G
2,4,5-Trichlorothiophenol ≥97.0% (by GC, titration analysis)
2,4,5-Trichlorothiophenol
分子式: C₆H₃Cl₃S
分子量: 213.51 g/mol
沸点: 154°C
MDL编号: MFCD00014447
CAS号: 3773-14-6
EINECS: 223-223-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f5b4c00d79e1790576b4ba343a7a84364823760&vid=77aa9f7a3d7cf38d19cb12eff3839c29ac6dd429&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 113.0 to 116.0 deg-C
Solubility in N,N-DMF almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR