2,5-二氯苯硫酚 ≥97.0% (通过GC)

供应商: TCI
Warning

D1145-25G D1145-5G
TCIAD1145-25GEA 0 CNY
TCIAD1145-25G TCIAD1145-5G
2,5-二氯苯硫酚 ≥97.0% (通过GC)
分子式: C₆H₄Cl₂S
沸点: 202 °C (1013 hPa)
溶点: 28 °C
密度: 1,415 g/cm³ (30 °C)
MDL编号: MFCD00004831
CAS号: 5858-18-4
EINECS: 227-486-9
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0756c30ede0b2bac0efc2857499d60931a233bce&vid=6309f5c26a5b3aecb12aaa0255250e9fe31014ba&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR