2-Chloro-5-nitroanisole ≥98.0% (通过GC)

供应商: TCI
Warning

C1773-25G C1773-5G
TCIAC1773-25GEA 0 CNY
TCIAC1773-25G TCIAC1773-5G
2-Chloro-5-nitroanisole ≥98.0% (通过GC)
分子式: C₇H₆ClNO₃
MDL编号: MFCD00079739
CAS号: 1009-36-5
EINECS: 213-768-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=665ac6e1efae1ffab53d77771e660d6b6bfe3bf4&vid=a146f6db3edaf306c82730fb568b514f6a608570&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR