2-氯苯胺 ≥98.0% (by GC)

供应商: TCI
Danger

C0111-25ML C0111-500ML
TCIAC0111-25MLEA 0 CNY
TCIAC0111-25ML TCIAC0111-500ML
2-氯苯胺 ≥98.0% (by GC)
2-氯苯胺
分子式: C₆H₆ClN
分子量: 127.57 g/mol
沸点: 209°C
溶点: –2°C
闪点: 104°C
存储条件: Ambient
MDL编号: MFCD00007656
CAS号: 95-51-2
EINECS: 202-426-4
UN号: 2019
ADR: 6.1 II
Merck Index: 14,02118
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f3039919a17e79c9aefda8183331cd4f346d3cd&vid=7ae9992ad1b57be7f65dd96ae53cccc7023078f1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR