4-氯-2-氟苯胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

C1169-10G C1169-25G
TCIAC1169-10GEA 0 CNY
TCIAC1169-10G TCIAC1169-25G
4-氯-2-氟苯胺 ≥98.0% (by GC, titration analysis)
4-氯-2-氟苯胺
分子式: C₆H₅ClFN
分子量: 145.56 g/mol
沸点: 92°C
闪点: 99°C
MDL编号: MFCD00010625
CAS号: 57946-56-2
EINECS: 261-034-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c038d250a556ea97075dc22a88052324485259dd&vid=5effc25faa3f84a5e82bdee76723e3e335a48b69&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR