3,5-二氟氯苯 ≥97.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Chloro-3,5-difluorobenzene

C2514-25G C2514-5G
TCIAC2514-25GEA 0 CNY
TCIAC2514-25G TCIAC2514-5G
3,5-二氟氯苯 ≥97.0% (by GC)
3,5-二氟氯苯
分子式: C₆H₃ClF₂
沸点: 116°C
密度: 1,329 g/cm³ (20 °C)
闪点: 29°C
MDL编号: MFCD00041518
CAS号: 1435-43-4
UN号: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c433bd057f874eda5e54289d1fbc1496417fd71&vid=40bc21b5fb89b98dc2945b47cc9d84da0a3c2a1c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR