1,3-二氯-2-氟苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,6-Dichlorofluorobenzene

D2485-25G D2485-5G
TCIAD2485-25GEA 0 CNY
TCIAD2485-25G TCIAD2485-5G
1,3-二氯-2-氟苯 ≥98.0% (by GC)
1,3-二氯-2-氟苯
分子式: C₆H₃Cl₂F
分子量: 164.99 g/mol
沸点: 171°C
溶点: 39
闪点: 60°C
存储条件: Refrigerator
MDL编号: MFCD00010307
CAS号: 2268-05-5
UN号: 1325
ADR: 4.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c1f375c74264fdaac9dacad422b0335d461ac21&vid=097ad33f1790cb5a1e09daff9950720341650b91&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 40.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR