4-氯-3,5-二硝基苯甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 3,5-Dinitro-4-chlorobenzoic acid

C0607-25G C0607-250G
TCIAC0607-25GEA 0 CNY
TCIAC0607-25G TCIAC0607-250G
4-氯-3,5-二硝基苯甲酸 ≥98.0% (by titrimetric analysis)
4-氯-3,5-二硝基苯甲酸
分子式: C₇H₃ClN₂O₆
分子量: 246.56 g/mol
沸点: 240
存储条件: Ambient
MDL编号: MFCD00007080
CAS号: 118-97-8
EINECS: 204-290-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dc9209195d4484884850976d5664d767658b30f&vid=2ac7b751d3ba24634e5398b877057ef18d0c7217&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 160.0 to 163.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR