4-氯-3-硝基苯甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
C0223-25G
C0223-500G
TCIAC0223-25GEA
0
CNY
TCIAC0223-25G
TCIAC0223-500G
4-氯-3-硝基苯甲酸 ≥98.0% (by GC, titration analysis)
4-氯-3-硝基苯甲酸
分子式:
C₇H₄ClNO₄ 分子量: 201.57 g/mol 存储条件: Ambient |
MDL编号:
MFCD00007079 CAS号: 96-99-1 EINECS: 202-550-9 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 181.0 to 185.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...