4-氨基-3,5-二氯苯甲腈 ≥98.0% (by GC)
供应商: TCI
Danger
A1928-1G A1928-5G
TCIAA1928-1GEA 0 CNY
TCIAA1928-1G TCIAA1928-5G
4-氨基-3,5-二氯苯甲腈 ≥98.0% (by GC)
4-氨基-3,5-二氯苯甲腈
分子式: C₇H₄Cl₂N₂
分子量: 187.03 g/mol
沸点: 323
溶点: 120
MDL编号: MFCD00017341
CAS号: 78473-00-4
EINECS: 278-914-6
UN号: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd9b838677a067f7c911e6e07e0418c22a3c5d6c&vid=af67f08e14ca2d482838281794edcf8cc1cc8f21&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C