3,5-二氯苯甲醛 ≥97.0% (通过GC)

供应商: TCI
Danger

D2012-1G D2012-5G
TCIAD2012-1GEA 0 CNY
TCIAD2012-1G TCIAD2012-5G
3,5-二氯苯甲醛 ≥97.0% (通过GC)
3,5-二氯苯甲醛
分子式: C₇H₄Cl₂O
沸点: 215°C
溶点: 63 °C
存储条件: Ambient
MDL编号: MFCD00003352
CAS号: 10203-08-4
EINECS: 233-499-0
UN号: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e9d1127adaf27a0ae49762701cded3ba1f671ad&vid=1fc931100f1173f6e9ab6df6e91460626fb5b224&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 63.0 to 66.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR