3,4-二氯苯甲醛 ≥98.0% (通过GC)

供应商: TCI
Danger

D0332-25G D0332-250G
TCIAD0332-25GEA 0 CNY
TCIAD0332-25G TCIAD0332-250G
3,4-二氯苯甲醛 ≥98.0% (通过GC)
3,4-二氯苯甲醛
分子式: C₇H₄Cl₂O
沸点: 248°C
溶点: 43 °C
存储条件: Refrigerator
MDL编号: MFCD00003351
CAS号: 6287-38-3
EINECS: 228-520-5
UN号: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d5c149caf1585a7fad8d75b75b6f83de8079258&vid=1a366c7cb548dcb8f3850c5d94131d9b4222439e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR