1,2,3,4-四氯苯 ≥96.0% (by GC)

供应商: TCI
Danger

T0631-25G T0631-500G
TCIAT0631-25GEA 0 CNY
TCIAT0631-25G TCIAT0631-500G
1,2,3,4-四氯苯 ≥96.0% (by GC)
1,2,3,4-四氯苯
分子式: C₆H₂Cl₄
分子量: 215.89 g/mol
沸点: 254°C
存储条件: Ambient
MDL编号: MFCD00000538
CAS号: 634-66-2
EINECS: 211-214-0
UN号: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=081430d9530cf3516d10ec875e0d4d4558a69189&vid=969b35f7e2b0c0de351f5f4a509c3cc14d44cae6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 45.0 to 47.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR