2,5-二羟基苯甲酸 ≥98.0% (通过HPLC,滴定分析)

供应商: TCI
Warning

D0569-25G D0569-500G
TCIAD0569-25GEA 0 CNY
TCIAD0569-25G TCIAD0569-500G
2,5-二羟基苯甲酸 ≥98.0% (通过HPLC,滴定分析)
2,5-二羟基苯甲酸
分子式: C₇H₆O₄
沸点: 339 °C (1013 hPa)
溶点: 202…206 °C
密度: 1,55 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00002460
CAS号: 490-79-9
EINECS: 207-718-5
Merck Index: 14,04398
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3be5e2c1fccc422476662fda5f1cf3feec5d3139&vid=579ffa6688289b130ef0722b01c0131d8034af22&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 204.0 to 208.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR