2,3-二羟基苯甲酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: Pyrocatechuic acid

C0054-25G C0054-5G
TCIAC0054-25GEA 0 CNY
TCIAC0054-25G TCIAC0054-5G
2,3-二羟基苯甲酸 ≥98.0% (by HPLC, titration analysis)
2,3-二羟基苯甲酸
分子式: C₇H₆O₄
分子量: 154.12 g/mol
沸点: 339
存储条件: Ambient
MDL编号: MFCD00002446
CAS号: 303-38-8
EINECS: 206-139-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eb19bc0bbed1ee5b2da78a002770a445926e208&vid=b590f884c26f63ab707844051a18de8e971d2566&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR