对羟基苯甲酸 ≥99.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
p-Hydroxybenzoic Acid
H0207-25G
H0207-500G
TCIAH0207-25GEA
0
CNY
TCIAH0207-25G
TCIAH0207-500G
对羟基苯甲酸 ≥99.0% (by GC, titration analysis)
对羟基苯甲酸
分子式:
C₇H₆O₃ 分子量: 138.12 g/mol 沸点: 336 存储条件: Ambient |
MDL编号:
MFCD00002547 CAS号: 99-96-7 EINECS: 202-804-9 Merck Index: 14,04812 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 214.0 to 219.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...