间羟基苯甲酸 ≥98.0% (by HPLC, titration analysis)

供应商: TCI
Warning

Synonyms: m-Hydroxybenzoic Acid

H0205-25G H0205-500G
TCIAH0205-25GEA 0 CNY
TCIAH0205-25G TCIAH0205-500G
间羟基苯甲酸 ≥98.0% (by HPLC, titration analysis)
间羟基苯甲酸
分子式: C₇H₆O₃
分子量: 138.12 g/mol
沸点: 346
溶点: 199
闪点: 220
存储条件: Ambient
MDL编号: MFCD00002506
CAS号: 99-06-9
EINECS: 202-726-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4eec922f740c72a08d3e351fbe3dac2e3cb65d44&vid=304e634b99665f4d21b6899aa385faef148db780&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 201.0 to 207.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR