水杨酸 ≥99.5% (通过滴定分析)
供应商: TCI
|
Danger
|
H0206-25G
H0206-500G
TCIAH0206-25GEA
0
CNY
TCIAH0206-25G
TCIAH0206-500G
水杨酸 ≥99.5% (通过滴定分析)
分子式:
(HO)C₆H₄CO₂H 沸点: 211°C 溶点: 157…159 °C 密度: 1,443 g/cm³ (20 °C) 闪点: 157 °C 存储条件: Ambient |
MDL编号:
MFCD00002439 CAS号: 69-72-7 EINECS: 200-712-3 Merck Index: 14,08332 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 99.5 % |
Melting point | 158.0 to 161.0 deg-C |
Solubility in EtOH (95vol%) | transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...