2-羟基-3-甲氧基苯甲酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-甲氧基水杨酸

M1153-25G M1153-5G
TCIAM1153-25GEA 0 CNY
TCIAM1153-25G TCIAM1153-5G
2-羟基-3-甲氧基苯甲酸 ≥98.0% (by GC, titration analysis)
2-羟基-3-甲氧基苯甲酸
分子式: C₈H₈O₄
分子量: 168.15 g/mol
存储条件: Ambient
MDL编号: MFCD00002445
CAS号: 877-22-5
EINECS: 212-888-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=741aa471b82e295a8a8e8940d1a50f4841b98d84&vid=8fce1686912e33a974f37d6648f0da834a2e803a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR