3-羟基-4-硝基苯甲酸 ≥96.0% (by GC, titration analysis)

供应商: TCI
Warning

H0635-25G H0635-250G
TCIAH0635-25GEA 0 CNY
TCIAH0635-25G TCIAH0635-250G
3-羟基-4-硝基苯甲酸 ≥96.0% (by GC, titration analysis)
3-羟基-4-硝基苯甲酸
分子式: C₇H₅NO₅
分子量: 183.12 g/mol
存储条件: Ambient
MDL编号: MFCD00007110
CAS号: 619-14-7
EINECS: 210-580-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df8de238bee1480cc4531525a168d61c191d4df3&vid=f87ad60b9219204ef7da3c87d5b9ae18d7786b5d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR