苯甲酸异丁酯 ≥99.0% (by GC)

供应商: TCI

Synonyms: Benzoic acid isobutyl ester

B0072-25G B0072-500G
TCIAB0072-25GEA 0 CNY
TCIAB0072-25G TCIAB0072-500G
苯甲酸异丁酯 ≥99.0% (by GC)
苯甲酸异丁酯
分子式: C₁₁H₁₄O₂
分子量: 178.23 g/mol
闪点: 120°C
MDL编号: MFCD00048344
CAS号: 120-50-3
EINECS: 204-401-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fb3e28f5e5b7d5fb03f4b6915e3f8dc5307bb84&vid=4d269d19b9b8ee183fc5bc512231bf901a70d93c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR