3,4-二羟基苯甲醛 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 原儿茶醛

D0566-25G D0566-250G
TCIAD0566-25GEA 0 CNY
TCIAD0566-25G TCIAD0566-250G
3,4-二羟基苯甲醛 ≥98.0% (by GC, titration analysis)
3,4-二羟基苯甲醛
分子式: C₇H₆O₃
分子量: 138.12 g/mol
沸点: 295
存储条件: Ambient
MDL编号: MFCD00003370
CAS号: 139-85-5
EINECS: 205-377-7
Merck Index: 14,07893
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8417e759632e6ebf07a73190c07bfb2c5cddae9&vid=f842dc85342ccdaa03115d1ada66369b013adc95&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR