2,4,5-三甲氧基苯甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 细辛醛

T1172-25G T1172-5G
TCIAT1172-25GEA 0 CNY
TCIAT1172-25G TCIAT1172-5G
2,4,5-三甲氧基苯甲醛 ≥98.0% (by GC)
2,4,5-三甲氧基苯甲醛
分子式: C₁₀H₁₂O₄
分子量: 196.2 g/mol
沸点: 335
溶点: 112
存储条件: Ambient
MDL编号: MFCD00003312
CAS号: 4460-86-0
EINECS: 224-713-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a68fd20cee8ed09d87a961f445830422c4a5c5fe&vid=c038cf523fa29b83f670a3196ef76f10593dde12&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR