间茴香醛 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
A0478-25ML
A0478-500ML
TCIAA0478-25MLEA
0
CNY
TCIAA0478-25ML
TCIAA0478-500ML
间茴香醛 ≥98.0% (通过GC)
间茴香醛
分子式:
C₈H₈O₂ 沸点: 230°C 密度: 1,1149 g/cm³ (20 °C) 闪点: 121°C 存储条件: Ambient |
MDL编号:
MFCD00003361 CAS号: 591-31-1 EINECS: 209-712-8 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...