3-溴苯酚 ≥95.0% (by GC)
供应商: TCI
Warning
B0629-25G B0629-500G
TCIAB0629-25GEA 0 CNY
TCIAB0629-25G TCIAB0629-500G
3-溴苯酚 ≥95.0% (by GC)
3-溴苯酚
分子式: C₆H₅BrO
分子量: 173.01 g/mol
沸点: 237°C
存储条件: Refrigerator
MDL编号: MFCD00002253
CAS号: 591-20-8
EINECS: 209-706-5
Merck Index: 14,01428
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49e1f46d845f93dbbf509dfd09f26b441a05967a&vid=e27c6fecbbca949478cbe8832a8ff8c38d40755d&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 95.0 %