2,6-二溴苯酚 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
1,3-Dibromo-2-hydroxybenzene
D3384-25G
D3384-5G
TCIAD3384-25GEA
0
CNY
TCIAD3384-25G
TCIAD3384-5G
2,6-二溴苯酚 ≥98.0% (by titrimetric analysis)
2,6-二溴苯酚
分子式:
C₆H₄Br₂O 分子量: 251.91 g/mol 沸点: 256°C 存储条件: Ambient |
MDL编号:
MFCD00002152 CAS号: 608-33-3 EINECS: 210-161-0 UN号: 2811 ADR: 6.1 III |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 54.0 to 58.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...