2-溴-5-氟苯酚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-Bromo-4-fluoro-2-hydroxybenzene

B2040-25G B2040-5G
TCIAB2040-25GEA 0 CNY
TCIAB2040-25G TCIAB2040-5G
2-溴-5-氟苯酚 ≥98.0% (by GC)
2-溴-5-氟苯酚
分子式: C₆H₄BrFO
分子量: 191 g/mol
闪点: 87°C
MDL编号: MFCD00040939
CAS号: 147460-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9fcc368d8b0d498ab291195b897257dfb74c4b4&vid=584d2553d8057509baf0b7fde20a23cf26baf584&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR