4-溴-2-氟苯酚 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Bromo-3-fluoro-4-hydroxybenzene
B1835-25G
B1835-5G
TCIAB1835-25GEA
0
CNY
TCIAB1835-25G
TCIAB1835-5G
4-溴-2-氟苯酚 ≥98.0% (by GC)
4-溴-2-氟苯酚
分子式:
C₆H₄BrFO 分子量: 191 g/mol 沸点: 90°C 闪点: 98°C |
MDL编号:
MFCD00011722 CAS号: 2105-94-4 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...