1-溴-3-(三氟甲氧基)苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
3-(Trifluoromethoxy)bromobenzene
B2007-25G
B2007-5G
TCIAB2007-25GEA
0
CNY
TCIAB2007-25G
TCIAB2007-5G
1-溴-3-(三氟甲氧基)苯 ≥98.0% (by GC)
1-溴-3-(三氟甲氧基)苯
分子式:
C₇H₄BrF₃O 分子量: 241.01 g/mol 沸点: 81°C 闪点: 62°C |
MDL编号:
MFCD00040943 CAS号: 2252-44-0 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...