溴五氟苯 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-Bromo-2,3,4,5,6-pentafluorobenzene
B1116-25G
B1116-5G
TCIAB1116-25GEA
0
CNY
TCIAB1116-25G
TCIAB1116-5G
溴五氟苯 ≥99.0% (by GC)
溴五氟苯
分子式:
C₆BrF₅ 分子量: 246.96 g/mol 沸点: 137°C 溶点: -31 闪点: 87°C 存储条件: Refrigerator |
MDL编号:
MFCD00000287 CAS号: 344-04-7 EINECS: 206-449-0 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...