2,4-二氯苯肼 ≥98.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
(2,4-Dichlorophenyl)hydrazine hydrochloride, 2,4-Dichlorophenylhydrazine hydrochloride, 1-(2,4-Dichlorophenyl)hydrazine hydrochloride
D2433-25G
D2433-5G
TCIAD2433-25GEA
0
CNY
TCIAD2433-25G
TCIAD2433-5G
2,4-二氯苯肼 ≥98.0% (by HPLC, titration analysis)
2,4-二氯苯肼
分子式:
C₆H₇Cl₃N₂ 分子量: 213.49 g/mol |
MDL编号:
MFCD00012929 CAS号: 5446-18-4 EINECS: 226-659-6 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Potassium iodate Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...