N-亚硝基二乙胺 ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: 二乙基亚硝胺

D0516-25ML D0516-5ML
TCIAD0516-25MLEA 0 CNY
TCIAD0516-25ML TCIAD0516-5ML
N-亚硝基二乙胺 ≥99.0% (by GC)
N-亚硝基二乙胺
分子式: C₄H₁₀N₂O
MDL编号: MFCD00013890
CAS号: 55-18-5
EINECS: 200-226-1
UN号: 2810
ADR: 6.1,III
Merck Index: 14,06636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fba5c54366a120843b607562299613b0381b1270&vid=fba5c54366a120843b607562299613b0381b1270&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR