邻甲基苯肼 ≥98.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 2-甲基苯肼

T0318-25G T0318-5G
TCIAT0318-25GEA 0 CNY
TCIAT0318-25G TCIAT0318-5G
邻甲基苯肼 ≥98.0% (by titrimetric analysis)
邻甲基苯肼
分子式: C₇H₁₁ClN₂
分子量: 158.63 g/mol
MDL编号: MFCD00012925
CAS号: 635-26-7
Merck Index: 14,09542
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=216eac52151fbc38ee429efbd7afa0c489dedfae&vid=216eac52151fbc38ee429efbd7afa0c489dedfae&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR