1,3-二溴四氟苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1,3-Dibromo-2,4,5,6-tetrafluorobenzene

D1492-1G D1492-5G
TCIAD1492-1GEA 0 CNY
TCIAD1492-1G TCIAD1492-5G
1,3-二溴四氟苯 ≥98.0% (by GC)
1,3-二溴四氟苯
分子式: C₆Br₂F₄
沸点: 85 °C (18 -rr)
MDL编号: MFCD00031533
CAS号: 1559-87-1
EINECS: 216-326-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=462e6683109750a208db6bd6dae8aa32e0e44e10&vid=6566717e601b471bf373a2d55b849b6408f7f233&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR